Assay Details
| Gene Symbol: cup-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027550.4 | NP_001022721.2 | 8 - 9 | 1728 | 148 | |
| NM_001027551.5 | NP_001022722.2 | 7 - 8 | 1668 | 148 | ||
| NM_001268016.4 | NP_001254945.1 | 7 - 8 | 1680 | 148 | ||
| NM_001382919.1 | 7 - 8 | 1677 | 148 | |||
| NM_066263.6 | NP_498664.3 | 8 - 9 | 1693 | 148 | ||
| GenBank mRNA | AF482952.1 | - | 7 - 8 | 1674 | 148 | |
More Information
Set Membership: |
|

cup-5
Genomic Map