Assay Details
| Gene Symbol: cup-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027551.5 | NP_001022722.2 | 8 - 9 | 1807 | 76 | |
| NM_001268016.4 | NP_001254945.1 | 8 - 9 | 1819 | 76 | ||
| GenBank mRNA | AF482952.1 | - | 8 - 9 | 1813 | 76 | |
More Information
Set Membership: |
|

cup-5
Genomic Map