Assay Details
| Gene Symbol: cup-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027551.5 | NP_001022722.2 | 9 - 10 | 1889 | 88 | |
| NM_001268016.4 | NP_001254945.1 | 9 - 10 | 1901 | 88 | ||
| NM_001382919.1 | 9 - 10 | 1902 | 88 | |||
| NM_066263.6 | NP_498664.3 | 9 - 10 | 2229 | 88 | ||
| GenBank mRNA | AF482952.1 | - | 9 - 10 | 1895 | 88 | |
More Information
Set Membership: |
|

cup-5
Genomic Map