Assay Details
| Gene Symbol: fbn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276780.3 | NP_001263709.1 | 4 - 5 | 622 | 82 | |
| NM_001276782.3 | NP_001263711.1 | 4 - 5 | 632 | 82 | ||
| NM_001276783.3 | NP_001263712.1 | 4 - 5 | 622 | 82 | ||
| NM_001276785.3 | NP_001263714.1 | 4 - 5 | 622 | 82 | ||
| NM_001276786.3 | NP_001263715.1 | 4 - 5 | 622 | 82 | ||
| NM_001276787.4 | NP_001263716.1 | 4 - 5 | 622 | 82 | ||
| NM_001276788.3 | NP_001263717.1 | 4 - 5 | 622 | 82 | ||
| NM_001276789.3 | NP_001263718.1 | 4 - 5 | 622 | 82 | ||
| NM_066269.6 | NP_498670.2 | 4 - 5 | 622 | 82 | ||
| GenBank mRNA | JQ990141.1 | - | 4 - 5 | 622 | 82 | |
More Information
Set Membership: |
|

fbn-1
Genomic Map