Assay Details
| Gene Symbol: ncl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312923.2 | NP_001299852.1 | 2 - 3 | 254 | 65 | |
| NM_001312924.2 | NP_001299853.1 | 2 - 3 | 188 | 65 | ||
| NM_001312995.3 | NP_001299924.1 | 1 - 2 | 152 | 65 | ||
| NM_001379801.1 | NP_001368538.1 | 1 - 2 | 152 | 65 | ||
| NM_001379802.1 | NP_001367412.1 | 1 - 2 | 169 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379802.1 transcripts.
Set Membership: |
|

ncl-1
Genomic Map