Assay Details
| Gene Symbol: ncl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312923.2 | NP_001299852.1 | 7 - 8 | 1330 | 74 | |
| NM_001312924.2 | NP_001299853.1 | 7 - 8 | 1273 | 74 | ||
| NM_001312995.3 | NP_001299924.1 | 6 - 7 | 1237 | 74 | ||
| NM_001312997.3 | NP_001299926.1 | 5 - 6 | 1075 | 74 | ||
| NM_001312998.3 | NP_001299927.1 | 5 - 6 | 1066 | 74 | ||
| NM_001312999.3 | NP_001299928.1 | 4 - 5 | 889 | 74 | ||
| NM_001379801.1 | NP_001368538.1 | 6 - 7 | 1228 | 74 | ||
| NM_001379802.1 | NP_001367412.1 | 6 - 7 | 1245 | 74 | ||
| GenBank mRNA | AF047027.1 | - | 4 - 5 | 1014 | 74 | |
More Information
Set Membership: |
|

ncl-1
Genomic Map