Assay Details
| Gene Symbol: ncl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312923.2 | NP_001299852.1 | 10 - 11 | 2547 | 87 | |
| NM_001312924.2 | NP_001299853.1 | 10 - 11 | 2490 | 87 | ||
| NM_001312995.3 | NP_001299924.1 | 9 - 10 | 2454 | 87 | ||
| NM_001312997.3 | NP_001299926.1 | 8 - 9 | 2292 | 87 | ||
| NM_001312998.3 | NP_001299927.1 | 8 - 9 | 2283 | 87 | ||
| NM_001312999.3 | NP_001299928.1 | 7 - 8 | 2106 | 87 | ||
| NM_001379801.1 | NP_001368538.1 | 9 - 10 | 2445 | 87 | ||
| NM_001379802.1 | NP_001367412.1 | 9 - 10 | 2462 | 87 | ||
| GenBank mRNA | AF047027.1 | - | 7 - 8 | 2231 | 87 | |
More Information
Set Membership: |
|

ncl-1
Genomic Map