Assay Details
| Gene Symbol: ncl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312923.2 | NP_001299852.1 | 11 - 12 | 2836 | 67 | |
| NM_001312924.2 | NP_001299853.1 | 11 - 12 | 2779 | 67 | ||
| NM_001312995.3 | NP_001299924.1 | 10 - 11 | 2743 | 67 | ||
| NM_001312997.3 | NP_001299926.1 | 9 - 10 | 2581 | 67 | ||
| NM_001312998.3 | NP_001299927.1 | 9 - 10 | 2572 | 67 | ||
| NM_001312999.3 | NP_001299928.1 | 8 - 9 | 2395 | 67 | ||
| NM_001379801.1 | NP_001368538.1 | 10 - 11 | 2734 | 67 | ||
| NM_001379802.1 | NP_001367412.1 | 10 - 11 | 2751 | 67 | ||
| GenBank mRNA | AF047027.1 | - | 8 - 9 | 2520 | 67 | |
More Information
Set Membership: |
|

ncl-1
Genomic Map