Assay Details
| Gene Symbol: cup-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066309.6 | NP_498710.2 | 2 - 3 | 388 | 129 | |
| GenBank mRNA | AY611497.1 | - | 2 - 3 | 368 | 129 | |
More Information
Set Membership: |
|

cup-4
Genomic Map