Assay Details
| Gene Symbol: cup-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066309.6 | NP_498710.2 | 3 - 4 | 497 | 76 | |
| GenBank mRNA | AY611497.1 | - | 3 - 4 | 477 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cup-4| Gene Symbol: cup-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066309.6 | NP_498710.2 | 3 - 4 | 497 | 76 | |
| GenBank mRNA | AY611497.1 | - | 3 - 4 | 477 | 76 | |
Set Membership: |
|