Assay Details
| Gene Symbol: pcp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379810.2 | NP_001367399.1 | 7 - 8 | 939 | 152 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pcp-5| Gene Symbol: pcp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379810.2 | NP_001367399.1 | 7 - 8 | 939 | 152 | |
Set Membership: |
|