Assay Details
| Gene Symbol: clp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379817.1 | NP_001367450.1 | 2 - 3 | 637 | 99 | |
| NM_066340.6 | NP_498741.3 | 4 - 5 | 534 | 99 | ||
More Information
Set Membership: |
|

clp-1
Genomic Map