Assay Details
| Gene Symbol: cit-1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382929.1 | 1 - 2 | 107 | 63 | ||
| NM_066344.5 | NP_498745.1 | 1 - 2 | 101 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382929.1 transcripts.
Set Membership: |
|

cit-1.2
Genomic Map