Assay Details
| Gene Symbol: pros-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066359.5 | NP_498760.2 | 8 - 9 | 1655 | 145 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pros-1| Gene Symbol: pros-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066359.5 | NP_498760.2 | 8 - 9 | 1655 | 145 | |
Set Membership: |
|