Assay Details
| Gene Symbol: dcr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066360.4 | NP_498761.2 | 13 - 14 | 2348 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dcr-1| Gene Symbol: dcr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066360.4 | NP_498761.2 | 13 - 14 | 2348 | 81 | |
Set Membership: |
|