Assay Details
| Gene Symbol: idi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382931.1 | 18 - 19 | 3818 | 145 | ||
| GenBank mRNA | AY597336.1 | - | 1 - 2 | 204 | 145 | |
| Gene Symbol: vms-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066364.7 | NP_498765.1 | 11 - 12 | 2278 | 145 | |
More Information
Set Membership: |
|

idi-1
Genomic Map