Assay Details
| Gene Symbol: idi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382931.1 | 20 - 21 | 4225 | 69 | ||
| GenBank mRNA | AY597336.1 | - | 3 - 4 | 611 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

idi-1| Gene Symbol: idi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382931.1 | 20 - 21 | 4225 | 69 | ||
| GenBank mRNA | AY597336.1 | - | 3 - 4 | 611 | 69 | |
Set Membership: |
|