Assay Details
| Gene Symbol: C14B9.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066374.9 | NP_498775.2 | 2 - 3 | 307 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C14B9.2| Gene Symbol: C14B9.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066374.9 | NP_498775.2 | 2 - 3 | 307 | 100 | |
Set Membership: |
|