Assay Details
| Gene Symbol: rpn-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066468.5 | NP_498869.1 | 3 - 4 | 1222 | 157 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpn-3| Gene Symbol: rpn-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066468.5 | NP_498869.1 | 3 - 4 | 1222 | 157 | |
Set Membership: |
|