Assay Details
| Gene Symbol: trp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047424.3 | NP_001040889.1 | 12 - 13 | 2414 | 98 | |
| NM_001379835.1 | NP_001367402.1 | 12 - 13 | 2426 | 98 | ||
| GenBank mRNA | AJ276027.1 | - | 12 - 13 | 2460 | 98 | |
More Information
Set Membership: |
|

trp-1
Genomic Map