Assay Details
| Gene Symbol: trp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047424.3 | NP_001040889.1 | 4 - 5 | 373 | 101 | |
| NM_001379835.1 | NP_001367402.1 | 4 - 5 | 385 | 101 | ||
| GenBank mRNA | AJ276027.1 | - | 4 - 5 | 419 | 101 | |
More Information
Set Membership: |
|

trp-1
Genomic Map