Assay Details
| Gene Symbol: C06E1.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066490.6 | NP_498891.2 | 5 - 6 | 494 | 152 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C06E1.7| Gene Symbol: C06E1.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066490.6 | NP_498891.2 | 5 - 6 | 494 | 152 | |
Set Membership: |
|