Assay Details
| Gene Symbol: rha-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066494.2 | NP_498895.1 | 10 - 11 | 2573 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rha-2| Gene Symbol: rha-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066494.2 | NP_498895.1 | 10 - 11 | 2573 | 123 | |
Set Membership: |
|