Assay Details
| Gene Symbol: dpy-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066508.2 | NP_498909.1 | 8 - 9 | 1352 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-19| Gene Symbol: dpy-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066508.2 | NP_498909.1 | 8 - 9 | 1352 | 123 | |
Set Membership: |
|