Assay Details
Gene Symbol: fli-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066512.6 | NP_498913.2 | 3 - 4 | 633 | 124 | |
GenBank mRNA | U01183.1 | - | 4 - 5 | 666 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
