Assay Details
Gene Symbol: fli-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066512.6 | NP_498913.2 | 5 - 6 | 1161 | 88 | |
GenBank mRNA | U01183.1 | - | 6 - 7 | 1194 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
