Assay Details
| Gene Symbol: tomm-70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066531.6 | NP_498932.3 | 9 - 10 | 1526 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tomm-70| Gene Symbol: tomm-70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066531.6 | NP_498932.3 | 9 - 10 | 1526 | 82 | |
Set Membership: |
|