Assay Details
| Gene Symbol: lin-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027845.5 | NP_001023016.1 | 3 - 4 | 319 | 80 | |
| GenBank mRNA | AF269693.1 | - | 3 - 4 | 322 | 80 | |
More Information
Set Membership: |
|

lin-9
Genomic Map