Assay Details
| Gene Symbol: lin-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027844.4 | NP_001023015.1 | 7 - 8 | 1597 | 141 | |
| NM_001027845.5 | NP_001023016.1 | 7 - 8 | 1594 | 141 | ||
| GenBank mRNA | AF269693.1 | - | 7 - 8 | 1597 | 141 | |
| AF269694.1 | - | 7 - 8 | 1591 | 141 | ||
More Information
Set Membership: |
|

lin-9
Genomic Map