Assay Details
| Gene Symbol: nac-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268047.2 | NP_001254976.1 | 11 - 12 | 1627 | 93 | |
| NM_001268048.2 | NP_001254977.1 | 10 - 11 | 1374 | 93 | ||
More Information
Set Membership: |
|

nac-2
Genomic Map