Assay Details
| Gene Symbol: nac-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268047.2 | NP_001254976.1 | 8 - 9 | 1168 | 155 | |
| NM_001268048.2 | NP_001254977.1 | 7 - 8 | 915 | 155 | ||
More Information
Set Membership: |
|

nac-2
Genomic Map