Assay Details
| Gene Symbol: mrpl-44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066611.6 | NP_499012.2 | 1 - 2 | 58 | 66 | |
| Gene Symbol: F02A9.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382956.1 | 1 - 2 | 58 | 66 | ||
More Information
Set Membership: |
|

mrpl-44
Genomic Map