Assay Details
| Gene Symbol: grl-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066619.2 | NP_499020.1 | 2 - 3 | 223 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

grl-11| Gene Symbol: grl-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066619.2 | NP_499020.1 | 2 - 3 | 223 | 134 | |
Set Membership: |
|