Assay Details
| Gene Symbol: nhl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322726.2 | NP_001309582.1 | 2 - 3 | 259 | 65 | |
| NM_066627.6 | NP_499028.1 | 2 - 3 | 259 | 65 | ||
More Information
Set Membership: |
|

nhl-1
Genomic Map