Assay Details
| Gene Symbol: dgk-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066630.4 | NP_499031.3 | 12 - 13 | 2129 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dgk-3| Gene Symbol: dgk-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066630.4 | NP_499031.3 | 12 - 13 | 2129 | 88 | |
Set Membership: |
|