Assay Details
| Gene Symbol: cpb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066650.6 | NP_499051.3 | 4 - 5 | 990 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpb-1| Gene Symbol: cpb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066650.6 | NP_499051.3 | 4 - 5 | 990 | 70 | |
Set Membership: |
|