Assay Details
| Gene Symbol: emb-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027491.7 | NP_001022662.2 | 3 - 4 | 440 | 62 | |
| NM_001027492.6 | NP_001022663.1 | 3 - 4 | 391 | 62 | ||
More Information
Set Membership: |
|

emb-9
Genomic Map