Assay Details
| Gene Symbol: rgr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372642.1 | NP_001359524.1 | 14 - 15 | 4248 | 60 | |
| NM_066666.5 | NP_499067.3 | 14 - 15 | 3940 | 60 | ||
More Information
Set Membership: |
|

rgr-1
Genomic Map