Assay Details
| Gene Symbol: rheb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066678.4 | NP_499079.1 | 1 - 2 | 96 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rheb-1| Gene Symbol: rheb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066678.4 | NP_499079.1 | 1 - 2 | 96 | 124 | |
Set Membership: |
|