Assay Details
| Gene Symbol: bre-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066686.7 | NP_499087.2 | 3 - 4 | 364 | 83 | |
| NR_132454.1 | 3 - 4 | 342 | 83 | |||
| GenBank mRNA | AY533305.1 | - | 2 - 3 | 249 | 83 | |
More Information
Set Membership: |
|

bre-3
Genomic Map