Assay Details
| Gene Symbol: ctr-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268071.3 | NP_001255000.1 | 3 - 4 | 1239 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ctr-9| Gene Symbol: ctr-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268071.3 | NP_001255000.1 | 3 - 4 | 1239 | 66 | |
Set Membership: |
|