Assay Details
| Gene Symbol: tbg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066730.8 | NP_499131.1 | 3 - 4 | 600 | 97 | |
| GenBank mRNA | AF287259.1 | - | 3 - 4 | 622 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tbg-1| Gene Symbol: tbg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066730.8 | NP_499131.1 | 3 - 4 | 600 | 97 | |
| GenBank mRNA | AF287259.1 | - | 3 - 4 | 622 | 97 | |
Set Membership: |
|