Assay Details
| Gene Symbol: ubc-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066732.6 | NP_499133.1 | 2 - 3 | 150 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ubc-7| Gene Symbol: ubc-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066732.6 | NP_499133.1 | 2 - 3 | 150 | 79 | |
Set Membership: |
|