Assay Details
| Gene Symbol: vps-53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268098.3 | NP_001255027.2 | 3 - 4 | 1310 | 61 | |
| NM_001268099.4 | NP_001255028.2 | 1 - 2 | 638 | 61 | ||
More Information
Set Membership: |
|

vps-53
Genomic Map