Assay Details
| Gene Symbol: mcm-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027840.5 | NP_001023011.1 | 3 - 4 | 811 | 67 | |
| GenBank mRNA | AF326940.1 | - | 3 - 4 | 810 | 67 | |
More Information
Set Membership: |
|

mcm-6
Genomic Map