Assay Details
| Gene Symbol: riok-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066772.8 | NP_499173.1 | 1 - 2 | 91 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

riok-3| Gene Symbol: riok-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066772.8 | NP_499173.1 | 1 - 2 | 91 | 84 | |
Set Membership: |
|