Assay Details
| Gene Symbol: panl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066776.8 | NP_499177.3 | 2 - 3 | 298 | 76 | |
| Gene Symbol: arl-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382971.1 | 1 - 2 | 116 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382971.1 transcripts.
Set Membership: |
|

panl-3
Genomic Map