Assay Details
| Gene Symbol: spop-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027593.2 | NP_001022764.1 | 4 - 5 | 700 | 69 | |
| NM_001027594.5 | NP_001022765.1 | 3 - 4 | 600 | 69 | ||
More Information
Set Membership: |
|

spop-1
Genomic Map