Assay Details
| Gene Symbol: vps-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027816.3 | NP_001022987.1 | 3 - 4 | 333 | 105 | |
| NM_001027817.4 | NP_001022988.1 | 3 - 4 | 357 | 105 | ||
More Information
Set Membership: |
|

vps-29
Genomic Map