Assay Details
| Gene Symbol: ttll-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027814.3 | NP_001022985.1 | 5 - 6 | 846 | 101 | |
| NM_001027815.6 | NP_001022986.1 | 4 - 5 | 743 | 101 | ||
More Information
Set Membership: |
|

ttll-4
Genomic Map